Assay Details
| Gene Symbol: LINC01431 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_109884.1 | 2 - 3 | 258 | 130 | ||
| GenBank mRNA | AI693606.1 | - | 1 - 2 | 130 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | nuclear transport factor 2 like export factor 1 | NXT1 |
MTR2, P15 |
-1.8kb | Anti-sense | Chr. 20 - 23350736 - 23354772 on NCBI Build GRCh38 |
| Downstream | GDNF inducible zinc finger protein 1 | GZF1 |
ZBTB23, ZNF336 |
4.5kb | Anti-sense | Chr. 20 - 23361585 - 23373062 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LINC01431
Genomic Map