Assay Details
| Gene Symbol: SLC35B3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_109914.1 | 3 - 4 | 1038 | 114 | ||
| GenBank mRNA | BX647208.1 | - | 3 - 4 | 972 | 114 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | EEF1E1-BLOC1S5 readthrough (NMD candidate) | EEF1E1-BLOC1S5 | EEF1E1-MUTED | -318.1kb | Sense | Chr. 6 - 8013567 - 8102595 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC100506207 | LOC100506207 | 7.6kb | Anti-sense | Chr. 6 - 8435623 - 8785445 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

SLC35B3
Genomic Map