| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013208.hg.1 |
ENST00000421034.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013208.hg.1 |
ENST00000421034.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013208.hg.1 |
ENST00000421034.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013208.hg.1 |
ENST00000434969.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013208.hg.1 |
ENST00000434969.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013208.hg.1 |
ENST00000434969.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013208.hg.1 |
ENST00000434969.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013208.hg.1 |
ENST00000434969.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013208.hg.1 |
NR_109968.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013208.hg.1 |
NR_109968.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013208.hg.1 |
NR_109968.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013208.hg.1 |
NR_109968.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013208.hg.1 |
NR_109968.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013208.hg.1 |
ENST00000421034.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013208.hg.1 |
ENST00000421034.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013208.hg.1 |
ENST00000421034.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013208.hg.1 |
ENST00000434969.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013208.hg.1 |
ENST00000434969.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013208.hg.1 |
ENST00000434969.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013208.hg.1 |
ENST00000434969.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013208.hg.1 |
ENST00000434969.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013208.hg.1 |
NR_109968.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013208.hg.1 |
NR_109968.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013208.hg.1 |
NR_109968.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013208.hg.1 |
NR_109968.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013208.hg.1 |
NR_109968.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300198476.hg.1 |
ENST00000434969.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300198476.hg.1 |
NR_109968.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300198476.hg.1 |
ENST00000434969.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300198476.hg.1 |
NR_109968.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300198476.hg.1 |
TC0300013208.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0300198475.hg.1 |
ENST00000434969.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300198475.hg.1 |
NR_109968.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300198475.hg.1 |
ENST00000434969.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300198475.hg.1 |
NR_109968.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300198475.hg.1 |
TC0300013208.hg.1 |
Clariom D TC |