Assay Details
| Gene Symbol: CSF1R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_109969.1 | 13 - 14 | 2183 | 81 | ||
| GenBank mRNA | AB208819.1 | - | 13 - 14 | 2133 | 81 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | HMG-box containing 3 | HMGXB3 |
HMGX3, SMF |
-4.4kb | Anti-sense | Chr. 5 - 150000606 - 150053143 on NCBI Build GRCh38 |
| Downstream | platelet derived growth factor receptor beta | PDGFRB |
CD140B, IBGC4, IMF1, JTK12, KOGS, PDGFR, PDGFR-1, PDGFR1, PENTT |
52.9kb | Sense | Chr. 5 - 150113839 - 150155884 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

CSF1R
Genomic Map