Assay Details
| Gene Symbol: NRSN2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_109990.1 | 1 - 2 | 113 | 97 | ||
| GenBank mRNA | BX104512.1 | - | 1 - 2 | 111 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2000008123.hg.1 | ENST00000414676.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008123.hg.1 | ENST00000414676.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008123.hg.1 | ENST00000414676.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008123.hg.1 | ENST00000414676.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008123.hg.1 | ENST00000414676.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008123.hg.1 | ENST00000414676.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008123.hg.1 | ENST00000414676.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008123.hg.1 | ENST00000414676.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008123.hg.1 | ENST00000414676.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008123.hg.1 | ENST00000414676.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008123.hg.1 | ENST00000442637.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008123.hg.1 | ENST00000442637.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008123.hg.1 | ENST00000442637.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008123.hg.1 | ENST00000442637.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008123.hg.1 | ENST00000442637.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008123.hg.1 | ENST00000442637.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008123.hg.1 | NR_109990.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008123.hg.1 | NR_109990.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008123.hg.1 | NR_109990.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008123.hg.1 | NR_109990.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008123.hg.1 | NR_109990.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008123.hg.1 | NR_109990.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000157021.hg.1 | ENST00000414676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000157021.hg.1 | ENST00000414676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000157021.hg.1 | NR_109990.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000157021.hg.1 | TC2000008123.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000157021.hg.1 | ENST00000414676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000157021.hg.1 | ENST00000414676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000157021.hg.1 | NR_109990.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000157015.hg.1 | ENST00000414676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000157015.hg.1 | ENST00000414676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000157015.hg.1 | ENST00000442637.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000157015.hg.1 | NR_109990.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000157015.hg.1 | TC2000008123.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000157015.hg.1 | ENST00000414676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000157015.hg.1 | ENST00000414676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000157015.hg.1 | ENST00000442637.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000157015.hg.1 | NR_109990.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000157016.hg.1 | ENST00000414676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000157016.hg.1 | ENST00000414676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000157016.hg.1 | ENST00000442637.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000157016.hg.1 | NR_109990.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000157016.hg.1 | TC2000008123.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000157016.hg.1 | ENST00000414676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000157016.hg.1 | ENST00000414676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000157016.hg.1 | ENST00000442637.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000157016.hg.1 | NR_109990.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000157019.hg.1 | ENST00000414676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000157019.hg.1 | ENST00000414676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000157019.hg.1 | ENST00000442637.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000157019.hg.1 | NR_109990.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000157019.hg.1 | TC2000008123.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000157019.hg.1 | ENST00000414676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000157019.hg.1 | ENST00000414676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000157019.hg.1 | ENST00000442637.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000157019.hg.1 | NR_109990.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008123.hg.1 | ENST00000414676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008123.hg.1 | ENST00000414676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008123.hg.1 | ENST00000414676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008123.hg.1 | ENST00000414676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008123.hg.1 | ENST00000414676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008123.hg.1 | ENST00000414676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008123.hg.1 | ENST00000414676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008123.hg.1 | ENST00000414676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008123.hg.1 | ENST00000414676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008123.hg.1 | ENST00000414676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008123.hg.1 | ENST00000442637.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008123.hg.1 | ENST00000442637.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008123.hg.1 | ENST00000442637.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008123.hg.1 | ENST00000442637.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008123.hg.1 | ENST00000442637.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008123.hg.1 | ENST00000442637.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008123.hg.1 | NR_109990.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008123.hg.1 | NR_109990.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008123.hg.1 | NR_109990.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008123.hg.1 | NR_109990.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008123.hg.1 | NR_109990.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008123.hg.1 | NR_109990.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000157011.hg.1 | NR_109990.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000157011.hg.1 | TC2000008123.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000157011.hg.1 | NR_109990.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000157013.hg.1 | ENST00000414676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000157013.hg.1 | ENST00000414676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000157013.hg.1 | ENST00000442637.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000157013.hg.1 | NR_109990.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000157013.hg.1 | TC2000008123.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000157013.hg.1 | ENST00000414676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000157013.hg.1 | ENST00000414676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000157013.hg.1 | ENST00000442637.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000157013.hg.1 | NR_109990.1 | Refseq |

NRSN2-AS1
Genomic Map