Assay Details
| Gene Symbol: LINC00211 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110012.1 | 1 - 2 | 126 | 73 | ||
| GenBank mRNA | BG430067.1 | - | 1 - 2 | 126 | 73 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | CDC42 effector protein 3 | CDC42EP3 |
BORG2, CEP3, UB1 |
-192.6kb | Sense | Chr. 2 - 37641882 - 37672535 on NCBI Build GRCh38 |
| Downstream | regulator of microtubule dynamics 2 | RMDN2 |
BLOCK18, FAM82A, FAM82A1, PRO34163, PYST9371, RMD-2, RMD2, RMD4 |
45.0kb | Anti-sense | Chr. 2 - 37920789 - 38068032 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | ENST00000418746.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | ENST00000418746.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | ENST00000418746.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | ENST00000446799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | ENST00000446799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | ENST00000446799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | ENST00000446799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | ENST00000446799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | ENST00000446799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | ENST00000446799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | ENST00000446799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | ENST00000446799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | ENST00000446799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | NR_110011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | NR_110011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | NR_110011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | NR_110011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | NR_110011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | NR_110011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | NR_110011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | NR_110011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | NR_110011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | NR_110011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | NR_110012.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | NR_110012.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | NR_110012.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | NR_110012.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | NR_110012.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | NR_110012.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | NR_110012.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012274.hg.1 | NR_110012.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | ENST00000413792.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | ENST00000418746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | ENST00000418746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | ENST00000418746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | ENST00000446799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | ENST00000446799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | ENST00000446799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | ENST00000446799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | ENST00000446799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | ENST00000446799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | ENST00000446799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | ENST00000446799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | ENST00000446799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | ENST00000446799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | NR_110011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | NR_110011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | NR_110011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | NR_110011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | NR_110011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | NR_110011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | NR_110011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | NR_110011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | NR_110011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | NR_110011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | NR_110012.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | NR_110012.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | NR_110012.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | NR_110012.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | NR_110012.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | NR_110012.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | NR_110012.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012274.hg.1 | NR_110012.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200189632.hg.1 | ENST00000446799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200189632.hg.1 | ENST00000446799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200189632.hg.1 | NR_110012.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200189632.hg.1 | ENST00000446799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200189632.hg.1 | ENST00000446799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200189632.hg.1 | NR_110012.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200189632.hg.1 | TC0200012274.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200189630.hg.1 | ENST00000418746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200189630.hg.1 | ENST00000446799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200189630.hg.1 | ENST00000446799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200189630.hg.1 | NR_110012.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200189630.hg.1 | ENST00000418746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200189630.hg.1 | ENST00000446799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200189630.hg.1 | ENST00000446799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200189630.hg.1 | NR_110012.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200189630.hg.1 | TC0200012274.hg.1 | Clariom D TC |

LINC00211
Genomic Map