Assay Details
| Gene Symbol: DKC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110021.1 | 2 - 3 | 867 | 145 | ||
| GenBank mRNA | KC954523.1 | - | 2 - 3 | 690 | 145 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | GRB2 associated binding protein 3 | GAB3 | -13.3kb | Anti-sense | Chr. X - 154675249 - 154751583 on NCBI Build GRCh38 | |
| Downstream |
small nucleolar RNA, H/ACA box 36A |
SNORA36A | ACA36 | 2.5kb | Sense | Chr. X - 154768528 - 154768659 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

DKC1
Genomic Map