| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012384.hg.1 |
ENST00000544240.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012384.hg.1 |
ENST00000544240.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012384.hg.1 |
ENST00000544240.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012384.hg.1 |
ENST00000544240.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012384.hg.1 |
ENST00000544240.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012384.hg.1 |
ENST00000545739.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012384.hg.1 |
ENST00000545739.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012384.hg.1 |
ENST00000545739.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012384.hg.1 |
ENST00000545739.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012384.hg.1 |
NR_110057.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012384.hg.1 |
NR_110057.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012384.hg.1 |
NR_110057.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012384.hg.1 |
NR_110057.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200196965.hg.1 |
ENST00000545739.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200196965.hg.1 |
NR_110057.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200196965.hg.1 |
ENST00000545739.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200196965.hg.1 |
NR_110057.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200196965.hg.1 |
TC1200012384.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1200012384.hg.1 |
ENST00000544240.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012384.hg.1 |
ENST00000544240.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012384.hg.1 |
ENST00000544240.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012384.hg.1 |
ENST00000544240.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012384.hg.1 |
ENST00000544240.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012384.hg.1 |
ENST00000545739.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012384.hg.1 |
ENST00000545739.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012384.hg.1 |
ENST00000545739.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012384.hg.1 |
ENST00000545739.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012384.hg.1 |
NR_110057.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012384.hg.1 |
NR_110057.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012384.hg.1 |
NR_110057.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012384.hg.1 |
NR_110057.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200196963.hg.1 |
ENST00000545739.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200196963.hg.1 |
NR_110057.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200196963.hg.1 |
ENST00000545739.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200196963.hg.1 |
NR_110057.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200196963.hg.1 |
TC1200012384.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1200196966.hg.1 |
ENST00000545739.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200196966.hg.1 |
NR_110057.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200196966.hg.1 |
ENST00000545739.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200196966.hg.1 |
NR_110057.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200196966.hg.1 |
TC1200012384.hg.1 |
Clariom D TC |