Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027413.hg.1 |
TC06002188.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027413.hg.1 |
ENST00000450575.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027413.hg.1 |
NR_027113.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027413.hg.1 |
NR_027114.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013428.hg.1 |
ENST00000450575.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013428.hg.1 |
ENST00000450575.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013428.hg.1 |
ENST00000450575.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013428.hg.1 |
ENST00000450575.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013428.hg.1 |
ENST00000450575.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013428.hg.1 |
ENST00000450575.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013428.hg.1 |
NR_027113.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013428.hg.1 |
NR_027113.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013428.hg.1 |
NR_027113.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013428.hg.1 |
NR_027113.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013428.hg.1 |
NR_027113.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013428.hg.1 |
NR_027113.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013428.hg.1 |
NR_027114.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013428.hg.1 |
NR_027114.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013428.hg.1 |
NR_027114.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013428.hg.1 |
NR_027114.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013428.hg.1 |
NR_027114.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013428.hg.1 |
NR_027114.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013428.hg.1 |
NR_027114.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013428.hg.1 |
NR_027114.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013428.hg.1 |
NR_027114.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013428.hg.1 |
NR_110148.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013428.hg.1 |
NR_110148.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013428.hg.1 |
NR_110148.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013428.hg.1 |
NR_110148.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013428.hg.1 |
NR_110148.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013428.hg.1 |
NR_110148.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013428.hg.1 |
NR_110148.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013428.hg.1 |
NR_110148.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013428.hg.1 |
NR_110149.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013428.hg.1 |
NR_110149.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013428.hg.1 |
NR_110149.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013428.hg.1 |
NR_110149.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013428.hg.1 |
NR_110149.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013428.hg.1 |
NR_110149.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013428.hg.1 |
NR_110149.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027416.hg.1 |
TC06002188.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027416.hg.1 |
ENST00000450575.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027416.hg.1 |
NR_027114.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027415.hg.1 |
TC06002188.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027415.hg.1 |
NR_027114.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027414.hg.1 |
TC06002188.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027414.hg.1 |
ENST00000450575.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027414.hg.1 |
NR_027114.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600192749.hg.1 |
NR_027114.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600192749.hg.1 |
NR_027114.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600192749.hg.1 |
TC0600013428.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600013428.hg.1 |
ENST00000450575.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013428.hg.1 |
ENST00000450575.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013428.hg.1 |
ENST00000450575.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013428.hg.1 |
ENST00000450575.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013428.hg.1 |
ENST00000450575.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013428.hg.1 |
ENST00000450575.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013428.hg.1 |
NR_027113.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013428.hg.1 |
NR_027113.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013428.hg.1 |
NR_027113.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013428.hg.1 |
NR_027113.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013428.hg.1 |
NR_027113.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013428.hg.1 |
NR_027113.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013428.hg.1 |
NR_027114.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013428.hg.1 |
NR_027114.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013428.hg.1 |
NR_027114.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013428.hg.1 |
NR_027114.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013428.hg.1 |
NR_027114.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013428.hg.1 |
NR_027114.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013428.hg.1 |
NR_027114.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013428.hg.1 |
NR_027114.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013428.hg.1 |
NR_027114.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013428.hg.1 |
NR_110148.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013428.hg.1 |
NR_110148.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013428.hg.1 |
NR_110148.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013428.hg.1 |
NR_110148.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013428.hg.1 |
NR_110148.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013428.hg.1 |
NR_110148.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013428.hg.1 |
NR_110148.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013428.hg.1 |
NR_110148.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013428.hg.1 |
NR_110149.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013428.hg.1 |
NR_110149.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013428.hg.1 |
NR_110149.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013428.hg.1 |
NR_110149.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013428.hg.1 |
NR_110149.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013428.hg.1 |
NR_110149.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013428.hg.1 |
NR_110149.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600192747.hg.1 |
ENST00000450575.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600192747.hg.1 |
NR_027113.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600192747.hg.1 |
NR_027114.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600192747.hg.1 |
NR_110148.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600192747.hg.1 |
NR_110149.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600192747.hg.1 |
ENST00000450575.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600192747.hg.1 |
NR_027113.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600192747.hg.1 |
NR_027114.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600192747.hg.1 |
NR_110148.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600192747.hg.1 |
NR_110149.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600192747.hg.1 |
TC0600013428.hg.1 |
Clariom D TC |