| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011664.hg.1 |
ENST00000412712.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011664.hg.1 |
ENST00000412712.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011664.hg.1 |
ENST00000412712.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011664.hg.1 |
ENST00000412712.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011664.hg.1 |
ENST00000412712.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011664.hg.1 |
ENST00000412712.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011664.hg.1 |
ENST00000412712.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011664.hg.1 |
ENST00000412712.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011664.hg.1 |
ENST00000418957.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011664.hg.1 |
ENST00000418957.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011664.hg.1 |
ENST00000418957.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011664.hg.1 |
ENST00000418957.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011664.hg.1 |
ENST00000421298.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011664.hg.1 |
ENST00000421298.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011664.hg.1 |
ENST00000433340.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011664.hg.1 |
ENST00000433340.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011664.hg.1 |
ENST00000433340.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011664.hg.1 |
ENST00000433592.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011664.hg.1 |
ENST00000433592.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011664.hg.1 |
ENST00000433592.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011664.hg.1 |
ENST00000433592.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011664.hg.1 |
ENST00000453900.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011664.hg.1 |
ENST00000453900.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011664.hg.1 |
ENST00000453900.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011664.hg.1 |
ENST00000453900.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011664.hg.1 |
ENST00000455965.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011664.hg.1 |
ENST00000455965.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011664.hg.1 |
ENST00000455965.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011664.hg.1 |
ENST00000455965.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011664.hg.1 |
ENST00000455965.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011664.hg.1 |
ENST00000455965.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011664.hg.1 |
ENST00000455965.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011664.hg.1 |
ENST00000455965.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011664.hg.1 |
ENST00000455965.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011664.hg.1 |
ENST00000455965.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011664.hg.1 |
ENST00000455965.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011664.hg.1 |
ENST00000455965.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011664.hg.1 |
NR_110153.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011664.hg.1 |
NR_110153.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011664.hg.1 |
NR_110154.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011664.hg.1 |
NR_110154.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011664.hg.1 |
NR_110154.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011664.hg.1 |
NR_110154.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011664.hg.1 |
NR_110154.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011664.hg.1 |
NR_110154.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200184680.hg.1 |
ENST00000453900.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200184680.hg.1 |
ENST00000455965.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200184680.hg.1 |
ENST00000455965.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200184680.hg.1 |
NR_110154.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200184680.hg.1 |
ENST00000453900.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200184680.hg.1 |
ENST00000455965.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200184680.hg.1 |
ENST00000455965.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200184680.hg.1 |
NR_110154.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200184680.hg.1 |
TC0200011664.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0200011664.hg.1 |
ENST00000412712.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011664.hg.1 |
ENST00000412712.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011664.hg.1 |
ENST00000412712.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011664.hg.1 |
ENST00000412712.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011664.hg.1 |
ENST00000412712.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011664.hg.1 |
ENST00000412712.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011664.hg.1 |
ENST00000412712.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011664.hg.1 |
ENST00000412712.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011664.hg.1 |
ENST00000418957.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011664.hg.1 |
ENST00000418957.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011664.hg.1 |
ENST00000418957.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011664.hg.1 |
ENST00000418957.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011664.hg.1 |
ENST00000421298.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011664.hg.1 |
ENST00000421298.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011664.hg.1 |
ENST00000433340.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011664.hg.1 |
ENST00000433340.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011664.hg.1 |
ENST00000433340.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011664.hg.1 |
ENST00000433592.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011664.hg.1 |
ENST00000433592.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011664.hg.1 |
ENST00000433592.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011664.hg.1 |
ENST00000433592.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011664.hg.1 |
ENST00000453900.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011664.hg.1 |
ENST00000453900.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011664.hg.1 |
ENST00000453900.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011664.hg.1 |
ENST00000453900.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011664.hg.1 |
ENST00000455965.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011664.hg.1 |
ENST00000455965.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011664.hg.1 |
ENST00000455965.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011664.hg.1 |
ENST00000455965.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011664.hg.1 |
ENST00000455965.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011664.hg.1 |
ENST00000455965.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011664.hg.1 |
ENST00000455965.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011664.hg.1 |
ENST00000455965.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011664.hg.1 |
ENST00000455965.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011664.hg.1 |
ENST00000455965.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011664.hg.1 |
ENST00000455965.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011664.hg.1 |
ENST00000455965.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011664.hg.1 |
NR_110153.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200011664.hg.1 |
NR_110153.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200011664.hg.1 |
NR_110154.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200011664.hg.1 |
NR_110154.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200011664.hg.1 |
NR_110154.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200011664.hg.1 |
NR_110154.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200011664.hg.1 |
NR_110154.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200011664.hg.1 |
NR_110154.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200184679.hg.1 |
ENST00000455965.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200184679.hg.1 |
ENST00000455965.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200184679.hg.1 |
NR_110154.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200184679.hg.1 |
ENST00000455965.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200184679.hg.1 |
ENST00000455965.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200184679.hg.1 |
NR_110154.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200184679.hg.1 |
TC0200011664.hg.1 |
Clariom D TC |