Assay Details
| Gene Symbol: LOC101929241 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110165.1 | 1 - 2 | 114 | 78 | ||
| GenBank mRNA | BC038465.1 | - | 1 - 2 | 60 | 78 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 618 | LINC00618 | C14orf63 | -513.4kb | Sense | Chr. 14 - 96943579 - 96945394 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC100129345 | LOC100129345 | 170.3kb | Anti-sense | Chr. 14 - 97632647 - 97686658 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC101929241
Genomic Map