Assay Details
Gene Symbol: LOC101927204 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110184.1 | 3 - 4 | 629 | 65 | ||
GenBank mRNA | AK124475.1 | - | 3 - 4 | 576 | 65 | |
BC028022.1 | - | 3 - 4 | 629 | 65 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | family with sequence similarity 111 member B | FAM111B |
CANP, POIKTMP |
-6.6kb | Anti-sense | Chr. 11 - 59107185 - 59127415 on NCBI Build GRCh38 |
Downstream | family with sequence similarity 111 member A | FAM111A |
GCLEB, KCS2 |
6.6kb | Anti-sense | Chr. 11 - 59142746 - 59155039 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000531708.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000531708.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000531708.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000531708.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000531708.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000531708.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000532845.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000532845.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000532845.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000532845.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000533220.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000533220.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000533220.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100182709.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100182709.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100182709.hg.1 | TC1100010959.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100182712.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182712.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182712.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100182712.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182712.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182712.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100182712.hg.1 | TC1100010959.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100182715.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182715.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182715.hg.1 | ENST00000531708.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182715.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100182715.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182715.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182715.hg.1 | ENST00000531708.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182715.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100182715.hg.1 | TC1100010959.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100182718.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182718.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182718.hg.1 | ENST00000531708.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182718.hg.1 | ENST00000532845.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182718.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100182718.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182718.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182718.hg.1 | ENST00000531708.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182718.hg.1 | ENST00000532845.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182718.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100182718.hg.1 | TC1100010959.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100182724.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182724.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182724.hg.1 | ENST00000533220.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182724.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100182724.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182724.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182724.hg.1 | ENST00000533220.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182724.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100182724.hg.1 | TC1100010959.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100182714.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182714.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182714.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100182714.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182714.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182714.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100182714.hg.1 | TC1100010959.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100182713.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182713.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182713.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100182713.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182713.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182713.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100182713.hg.1 | TC1100010959.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000531708.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000531708.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000531708.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000531708.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000531708.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000531708.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000532845.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000532845.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000532845.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000532845.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000533220.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000533220.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000533220.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100182717.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182717.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182717.hg.1 | ENST00000531708.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182717.hg.1 | ENST00000532845.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182717.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100182717.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182717.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182717.hg.1 | ENST00000531708.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182717.hg.1 | ENST00000532845.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182717.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100182717.hg.1 | TC1100010959.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100182726.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100182726.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100182726.hg.1 | TC1100010959.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100182711.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182711.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182711.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100182711.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182711.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182711.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100182711.hg.1 | TC1100010959.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100182721.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182721.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182721.hg.1 | ENST00000531708.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182721.hg.1 | ENST00000532845.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182721.hg.1 | ENST00000533220.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182721.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100182721.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182721.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182721.hg.1 | ENST00000531708.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182721.hg.1 | ENST00000532845.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182721.hg.1 | ENST00000533220.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182721.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100182721.hg.1 | TC1100010959.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100182723.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182723.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182723.hg.1 | ENST00000531708.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182723.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100182723.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182723.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182723.hg.1 | ENST00000531708.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182723.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100182723.hg.1 | TC1100010959.hg.1 | Clariom D TC |
