Assay Details
| Gene Symbol: MIR3681HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110197.1 | 2 - 3 | 210 | 68 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 4262 | MIR4262 | -170.8kb | Anti-sense | Chr. 2 - 11836933 - 11836986 on NCBI Build GRCh38 | |
| Downstream | microRNA 3681 | MIR3681 | 92.2kb | Sense | Chr. 2 - 12199130 - 12199201 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200016407.hg.1 | ENST00000412294.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016407.hg.1 | ENST00000412294.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016407.hg.1 | ENST00000412294.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016407.hg.1 | ENST00000412294.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016407.hg.1 | ENST00000412294.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016407.hg.1 | ENST00000412294.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016407.hg.1 | ENST00000412294.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016407.hg.1 | ENST00000412294.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016407.hg.1 | ENST00000412294.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016407.hg.1 | ENST00000412294.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016407.hg.1 | ENST00000412294.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016407.hg.1 | ENST00000412294.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016407.hg.1 | ENST00000412294.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016407.hg.1 | ENST00000412294.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016407.hg.1 | ENST00000412294.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016407.hg.1 | ENST00000412294.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016407.hg.1 | ENST00000412294.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016407.hg.1 | ENST00000412294.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016407.hg.1 | ENST00000412606.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016407.hg.1 | ENST00000412606.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016407.hg.1 | ENST00000412606.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016407.hg.1 | ENST00000412606.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016407.hg.1 | ENST00000412606.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016407.hg.1 | ENST00000449986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016407.hg.1 | ENST00000449986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016407.hg.1 | ENST00000449986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016407.hg.1 | ENST00000450916.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016407.hg.1 | ENST00000450916.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016407.hg.1 | ENST00000450916.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016407.hg.1 | ENST00000451644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016407.hg.1 | ENST00000451644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016407.hg.1 | ENST00000451644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016407.hg.1 | ENST00000451644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016407.hg.1 | ENST00000451644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016407.hg.1 | ENST00000451644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016407.hg.1 | ENST00000451644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016407.hg.1 | ENST00000451644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016407.hg.1 | ENST00000451644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016407.hg.1 | ENST00000451644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016407.hg.1 | ENST00000451644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016407.hg.1 | ENST00000451644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016407.hg.1 | NR_110196.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016407.hg.1 | NR_110196.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016407.hg.1 | NR_110196.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016407.hg.1 | NR_110196.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016407.hg.1 | NR_110196.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016407.hg.1 | NR_110196.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016407.hg.1 | NR_110196.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016407.hg.1 | NR_110196.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016407.hg.1 | NR_110196.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016407.hg.1 | NR_110197.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016407.hg.1 | NR_110197.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016407.hg.1 | NR_110197.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016407.hg.1 | NR_110197.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016407.hg.1 | NR_110197.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016407.hg.1 | NR_110197.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016407.hg.1 | NR_110198.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016407.hg.1 | NR_110198.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016407.hg.1 | NR_110198.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016407.hg.1 | NR_110198.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016407.hg.1 | ENST00000412294.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016407.hg.1 | ENST00000412294.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016407.hg.1 | ENST00000412294.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016407.hg.1 | ENST00000412294.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016407.hg.1 | ENST00000412294.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016407.hg.1 | ENST00000412294.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016407.hg.1 | ENST00000412294.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016407.hg.1 | ENST00000412294.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016407.hg.1 | ENST00000412294.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016407.hg.1 | ENST00000412294.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016407.hg.1 | ENST00000412294.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016407.hg.1 | ENST00000412294.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016407.hg.1 | ENST00000412294.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016407.hg.1 | ENST00000412294.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016407.hg.1 | ENST00000412294.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016407.hg.1 | ENST00000412294.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016407.hg.1 | ENST00000412294.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016407.hg.1 | ENST00000412294.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016407.hg.1 | ENST00000412606.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016407.hg.1 | ENST00000412606.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016407.hg.1 | ENST00000412606.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016407.hg.1 | ENST00000412606.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016407.hg.1 | ENST00000412606.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016407.hg.1 | ENST00000449986.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016407.hg.1 | ENST00000449986.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016407.hg.1 | ENST00000449986.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016407.hg.1 | ENST00000450916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016407.hg.1 | ENST00000450916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016407.hg.1 | ENST00000450916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016407.hg.1 | ENST00000451644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016407.hg.1 | ENST00000451644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016407.hg.1 | ENST00000451644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016407.hg.1 | ENST00000451644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016407.hg.1 | ENST00000451644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016407.hg.1 | ENST00000451644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016407.hg.1 | ENST00000451644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016407.hg.1 | ENST00000451644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016407.hg.1 | ENST00000451644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016407.hg.1 | ENST00000451644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016407.hg.1 | ENST00000451644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016407.hg.1 | ENST00000451644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016407.hg.1 | NR_110196.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016407.hg.1 | NR_110196.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016407.hg.1 | NR_110196.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016407.hg.1 | NR_110196.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016407.hg.1 | NR_110196.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016407.hg.1 | NR_110196.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016407.hg.1 | NR_110196.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016407.hg.1 | NR_110196.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016407.hg.1 | NR_110196.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016407.hg.1 | NR_110197.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016407.hg.1 | NR_110197.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016407.hg.1 | NR_110197.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016407.hg.1 | NR_110197.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016407.hg.1 | NR_110197.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016407.hg.1 | NR_110197.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016407.hg.1 | NR_110198.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016407.hg.1 | NR_110198.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016407.hg.1 | NR_110198.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016407.hg.1 | NR_110198.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200221041.hg.1 | ENST00000449986.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221041.hg.1 | ENST00000451644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221041.hg.1 | ENST00000451644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221041.hg.1 | NR_110197.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200221041.hg.1 | ENST00000449986.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221041.hg.1 | ENST00000451644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221041.hg.1 | ENST00000451644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221041.hg.1 | NR_110197.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200221041.hg.1 | TC0200016407.hg.1 | Clariom D TC |

MIR3681HG
Genomic Map