Assay Details
| Gene Symbol: LOC101927043 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110207.1 | 1 - 2 | 181 | 112 | ||
| GenBank mRNA | AK094343.1 | - | 1 - 2 | 73 | 112 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | calmodulin 2 | CALM2 |
CAMII, LQT15, PHKD, PHKD2, caM |
-44.2kb | Sense | Chr. 2 - 47160082 - 47176936 on NCBI Build GRCh38 |
| Downstream | epithelial cell adhesion molecule | EPCAM |
DIAR5, EGP-2, EGP314, EGP40, ESA, HNPCC8, KS1/4, KSA, M4S1, MIC18, MK-1, TACSTD1, TROP1 |
24.1kb | Anti-sense | Chr. 2 - 47369148 - 47387028 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000413185.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000413185.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000413185.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000413185.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000413185.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000413185.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000419035.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000419035.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000419035.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000448713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000448713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000448713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000448713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000448713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000448713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000448713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000448713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000450550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000450550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000450550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000450550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000450550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000450550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000450550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000450550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | NR_110207.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | NR_110207.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | NR_110207.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | NR_110207.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | NR_110208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | NR_110208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | NR_110208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | NR_110208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | NR_110208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | NR_110208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | NR_110208.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200191302.hg.1 | ENST00000419035.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191302.hg.1 | NR_110207.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200191302.hg.1 | ENST00000419035.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191302.hg.1 | NR_110207.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200191302.hg.1 | TC0200012505.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200191320.hg.1 | ENST00000413185.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191320.hg.1 | ENST00000419035.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191320.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191320.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191320.hg.1 | ENST00000448713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191320.hg.1 | ENST00000448713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191320.hg.1 | ENST00000450550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191320.hg.1 | ENST00000450550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191320.hg.1 | NR_110207.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200191320.hg.1 | NR_110208.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200191320.hg.1 | ENST00000413185.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191320.hg.1 | ENST00000419035.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191320.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191320.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191320.hg.1 | ENST00000448713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191320.hg.1 | ENST00000448713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191320.hg.1 | ENST00000450550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191320.hg.1 | ENST00000450550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191320.hg.1 | NR_110207.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200191320.hg.1 | NR_110208.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200191320.hg.1 | TC0200012505.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200191303.hg.1 | ENST00000419035.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191303.hg.1 | NR_110207.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200191303.hg.1 | ENST00000419035.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191303.hg.1 | NR_110207.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200191303.hg.1 | TC0200012505.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200191324.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191324.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191324.hg.1 | NR_110207.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200191324.hg.1 | NR_110208.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200191324.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191324.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191324.hg.1 | NR_110207.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200191324.hg.1 | NR_110208.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200191324.hg.1 | TC0200012505.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000413185.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000413185.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000413185.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000413185.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000413185.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000413185.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000419035.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000419035.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000419035.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000448713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000448713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000448713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000448713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000448713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000448713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000448713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000448713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000450550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000450550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000450550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000450550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000450550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000450550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000450550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000450550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | NR_110207.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | NR_110207.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | NR_110207.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | NR_110207.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | NR_110208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | NR_110208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | NR_110208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | NR_110208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | NR_110208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | NR_110208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | NR_110208.1 | Refseq |

LOC101927043
Genomic Map