| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015161.hg.1 |
ENST00000421998.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015161.hg.1 |
ENST00000421998.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015161.hg.1 |
ENST00000421998.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015161.hg.1 |
ENST00000421998.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015161.hg.1 |
ENST00000421998.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015161.hg.1 |
ENST00000421998.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015161.hg.1 |
ENST00000421998.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015161.hg.1 |
ENST00000421998.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015161.hg.1 |
ENST00000421998.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015161.hg.1 |
ENST00000421998.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015161.hg.1 |
ENST00000427269.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015161.hg.1 |
ENST00000427269.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015161.hg.1 |
ENST00000427269.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015161.hg.1 |
ENST00000437717.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015161.hg.1 |
ENST00000437717.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015161.hg.1 |
ENST00000437717.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015161.hg.1 |
NR_110214.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015161.hg.1 |
NR_110214.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015161.hg.1 |
NR_110214.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015161.hg.1 |
NR_110214.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015161.hg.1 |
NR_110214.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015161.hg.1 |
NR_110214.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015161.hg.1 |
NR_110215.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015161.hg.1 |
NR_110215.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015161.hg.1 |
NR_110215.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015161.hg.1 |
NR_110215.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015161.hg.1 |
NR_110215.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015161.hg.1 |
NR_110216.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015161.hg.1 |
NR_110216.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015161.hg.1 |
NR_110216.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015161.hg.1 |
NR_110216.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015161.hg.1 |
NR_110216.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015161.hg.1 |
NR_110217.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015161.hg.1 |
NR_110217.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015161.hg.1 |
NR_110217.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015161.hg.1 |
NR_110217.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200211054.hg.1 |
ENST00000421998.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200211054.hg.1 |
ENST00000421998.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200211054.hg.1 |
ENST00000427269.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200211054.hg.1 |
NR_110214.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200211054.hg.1 |
NR_110215.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200211054.hg.1 |
NR_110216.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200211054.hg.1 |
NR_110217.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200211054.hg.1 |
ENST00000421998.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200211054.hg.1 |
ENST00000421998.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200211054.hg.1 |
ENST00000427269.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200211054.hg.1 |
NR_110214.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200211054.hg.1 |
NR_110215.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200211054.hg.1 |
NR_110216.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200211054.hg.1 |
NR_110217.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200211054.hg.1 |
TC0200015161.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0200211065.hg.1 |
NR_110214.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200211065.hg.1 |
NR_110214.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200211065.hg.1 |
TC0200015161.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0200211064.hg.1 |
ENST00000437717.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200211064.hg.1 |
NR_110214.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200211064.hg.1 |
ENST00000437717.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200211064.hg.1 |
NR_110214.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200211064.hg.1 |
TC0200015161.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0200211062.hg.1 |
ENST00000421998.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200211062.hg.1 |
ENST00000421998.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200211062.hg.1 |
NR_110215.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200211062.hg.1 |
NR_110216.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200211062.hg.1 |
ENST00000421998.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200211062.hg.1 |
ENST00000421998.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200211062.hg.1 |
NR_110215.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200211062.hg.1 |
NR_110216.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200211062.hg.1 |
TC0200015161.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0200015161.hg.1 |
ENST00000421998.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015161.hg.1 |
ENST00000421998.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015161.hg.1 |
ENST00000421998.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015161.hg.1 |
ENST00000421998.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015161.hg.1 |
ENST00000421998.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015161.hg.1 |
ENST00000421998.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015161.hg.1 |
ENST00000421998.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015161.hg.1 |
ENST00000421998.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015161.hg.1 |
ENST00000421998.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015161.hg.1 |
ENST00000421998.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015161.hg.1 |
ENST00000427269.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015161.hg.1 |
ENST00000427269.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015161.hg.1 |
ENST00000427269.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015161.hg.1 |
ENST00000437717.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015161.hg.1 |
ENST00000437717.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015161.hg.1 |
ENST00000437717.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015161.hg.1 |
NR_110214.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015161.hg.1 |
NR_110214.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015161.hg.1 |
NR_110214.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015161.hg.1 |
NR_110214.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015161.hg.1 |
NR_110214.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015161.hg.1 |
NR_110214.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015161.hg.1 |
NR_110215.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015161.hg.1 |
NR_110215.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015161.hg.1 |
NR_110215.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015161.hg.1 |
NR_110215.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015161.hg.1 |
NR_110215.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015161.hg.1 |
NR_110216.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015161.hg.1 |
NR_110216.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015161.hg.1 |
NR_110216.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015161.hg.1 |
NR_110216.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015161.hg.1 |
NR_110216.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015161.hg.1 |
NR_110217.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015161.hg.1 |
NR_110217.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015161.hg.1 |
NR_110217.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015161.hg.1 |
NR_110217.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200211058.hg.1 |
ENST00000421998.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200211058.hg.1 |
ENST00000421998.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200211058.hg.1 |
ENST00000437717.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200211058.hg.1 |
NR_110214.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200211058.hg.1 |
NR_110215.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200211058.hg.1 |
NR_110216.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200211058.hg.1 |
ENST00000421998.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200211058.hg.1 |
ENST00000421998.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200211058.hg.1 |
ENST00000437717.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200211058.hg.1 |
NR_110214.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200211058.hg.1 |
NR_110215.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200211058.hg.1 |
NR_110216.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200211058.hg.1 |
TC0200015161.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0200211053.hg.1 |
ENST00000421998.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200211053.hg.1 |
ENST00000421998.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200211053.hg.1 |
NR_110214.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200211053.hg.1 |
NR_110215.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200211053.hg.1 |
NR_110216.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200211053.hg.1 |
NR_110217.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200211053.hg.1 |
ENST00000421998.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200211053.hg.1 |
ENST00000421998.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200211053.hg.1 |
NR_110214.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200211053.hg.1 |
NR_110215.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200211053.hg.1 |
NR_110216.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200211053.hg.1 |
NR_110217.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200211053.hg.1 |
TC0200015161.hg.1 |
Clariom D TC |