Assay Details
| Gene Symbol: LOC101927196 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110217.1 | 1 - 2 | 98 | 133 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC105373782 | LOC105373782 | -562.2kb | Sense | Chr. 2 - 185164954 - 185167072 on NCBI Build GRCh38 | |
| Downstream | long intergenic non-protein coding RNA 1473 | LINC01473 | 293.1kb | Sense | Chr. 2 - 186033531 - 186086317 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200015161.hg.1 | ENST00000421998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015161.hg.1 | ENST00000421998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015161.hg.1 | ENST00000421998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015161.hg.1 | ENST00000421998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015161.hg.1 | ENST00000421998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015161.hg.1 | ENST00000421998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015161.hg.1 | ENST00000421998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015161.hg.1 | ENST00000421998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015161.hg.1 | ENST00000421998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015161.hg.1 | ENST00000421998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015161.hg.1 | ENST00000427269.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015161.hg.1 | ENST00000427269.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015161.hg.1 | ENST00000427269.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015161.hg.1 | ENST00000437717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015161.hg.1 | ENST00000437717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015161.hg.1 | ENST00000437717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015161.hg.1 | NR_110214.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015161.hg.1 | NR_110214.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015161.hg.1 | NR_110214.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015161.hg.1 | NR_110214.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015161.hg.1 | NR_110214.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015161.hg.1 | NR_110214.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015161.hg.1 | NR_110215.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015161.hg.1 | NR_110215.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015161.hg.1 | NR_110215.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015161.hg.1 | NR_110215.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015161.hg.1 | NR_110215.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015161.hg.1 | NR_110216.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015161.hg.1 | NR_110216.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015161.hg.1 | NR_110216.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015161.hg.1 | NR_110216.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015161.hg.1 | NR_110216.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015161.hg.1 | NR_110217.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015161.hg.1 | NR_110217.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015161.hg.1 | NR_110217.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015161.hg.1 | NR_110217.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200211057.hg.1 | ENST00000421998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200211057.hg.1 | ENST00000421998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200211057.hg.1 | ENST00000427269.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200211057.hg.1 | NR_110214.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200211057.hg.1 | NR_110215.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200211057.hg.1 | NR_110216.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200211057.hg.1 | NR_110217.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200211057.hg.1 | ENST00000421998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200211057.hg.1 | ENST00000421998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200211057.hg.1 | ENST00000427269.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200211057.hg.1 | NR_110214.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200211057.hg.1 | NR_110215.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200211057.hg.1 | NR_110216.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200211057.hg.1 | NR_110217.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200211057.hg.1 | TC0200015161.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200211067.hg.1 | ENST00000427269.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200211067.hg.1 | NR_110217.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200211067.hg.1 | ENST00000427269.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200211067.hg.1 | NR_110217.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200211067.hg.1 | TC0200015161.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200015161.hg.1 | ENST00000421998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015161.hg.1 | ENST00000421998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015161.hg.1 | ENST00000421998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015161.hg.1 | ENST00000421998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015161.hg.1 | ENST00000421998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015161.hg.1 | ENST00000421998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015161.hg.1 | ENST00000421998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015161.hg.1 | ENST00000421998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015161.hg.1 | ENST00000421998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015161.hg.1 | ENST00000421998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015161.hg.1 | ENST00000427269.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015161.hg.1 | ENST00000427269.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015161.hg.1 | ENST00000427269.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015161.hg.1 | ENST00000437717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015161.hg.1 | ENST00000437717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015161.hg.1 | ENST00000437717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015161.hg.1 | NR_110214.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015161.hg.1 | NR_110214.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015161.hg.1 | NR_110214.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015161.hg.1 | NR_110214.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015161.hg.1 | NR_110214.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015161.hg.1 | NR_110214.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015161.hg.1 | NR_110215.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015161.hg.1 | NR_110215.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015161.hg.1 | NR_110215.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015161.hg.1 | NR_110215.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015161.hg.1 | NR_110215.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015161.hg.1 | NR_110216.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015161.hg.1 | NR_110216.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015161.hg.1 | NR_110216.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015161.hg.1 | NR_110216.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015161.hg.1 | NR_110216.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015161.hg.1 | NR_110217.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015161.hg.1 | NR_110217.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015161.hg.1 | NR_110217.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015161.hg.1 | NR_110217.1 | Refseq |

LOC101927196
Genomic Map