Assay Details
| Gene Symbol: LINC01250 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110228.1 | 3 - 4 | 438 | 117 | ||
| GenBank mRNA | AK095310.1 | - | 3 - 4 | 438 | 117 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | DB477601.1 | - | 107 | Chr. - - [-] on Build | 117 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101927534 | LOC101927534 | -95.3kb | Sense | Chr. 2 - 2870558 - 2871231 on NCBI Build GRCh38 | |
| Downstream | tumor suppressing subtransferable candidate 1 | TSSC1 | 63.5kb | Sense | Chr. 2 - 3188968 - 3377882 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

LINC01250
Genomic Map