Assay Details
| Gene Symbol: LOC101928371 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110236.1 | 1 - 2 | 534 | 145 | ||
| GenBank mRNA | BC044304.1 | - | 1 - 2 | 534 | 145 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | testis expressed 37 | TEX37 |
C2orf51, TSC21 |
-9.1kb | Sense | Chr. 2 - 88524623 - 88529585 on NCBI Build GRCh38 |
| Downstream | eukaryotic translation initiation factor 2 alpha kinase 3 | EIF2AK3 |
PEK, PERK, WRS |
16.6kb | Anti-sense | Chr. 2 - 88556740 - 88627576 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC101928371
Genomic Map