Assay Details
Gene Symbol: LOC101929596 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110259.1 | 1 - 2 | 295 | 66 | ||
GenBank mRNA | BU172790.1 | - | 1 - 2 | 295 | 66 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | atlastin GTPase 2 | ATL2 |
ARL3IP2, ARL6IP2, atlastin2 |
-118.1kb | Sense | Chr. 2 - 38293957 - 38378579 on NCBI Build GRCh38 |
Downstream | heterogeneous nuclear ribonucleoprotein L like | HNRNPLL |
HNRPLL, SRRF |
46.2kb | Sense | Chr. 2 - 38563186 - 38603036 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
