Assay Details
| Gene Symbol: LOC101929723 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110262.1 | 2 - 3 | 564 | 116 | ||
| GenBank mRNA | CA448932.1 | - | 2 - 3 | 116 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC388942 | LOC388942 | -38.1kb | Anti-sense | Chr. 2 - 41877555 - 41894046 on NCBI Build GRCh38 | |
| Downstream | chromosome 2 open reading frame 91 | C2orf91 | 2.8kb | Sense | Chr. 2 - 41935368 - 41961382 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC101929723
Genomic Map