Assay Details
| Gene Symbol: LOC101927948 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110287.1 | 1 - 2 | 65 | 78 | ||
| GenBank mRNA | BC042517.1 | - | 1 - 2 | 65 | 78 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101927967 | LOC101927967 | Anti-sense | Chr. 2 - 77743696 - 78290731 on NCBI Build GRCh38 | ||
| Downstream | uncharacterized LOC105374820 | LOC105374820 | 448.5kb | Sense | Chr. 2 - 78545531 - 78566088 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC101927948
Genomic Map