Assay Details
| Gene Symbol: GPATCH2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110314.1 | 7 - 8 | 1217 | 89 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | intraflagellar transport 43 | IFT43 |
C14orf179, CED3 |
-94.1kb | Sense | Chr. 14 - 75985753 - 76084073 on NCBI Build GRCh38 |
| Downstream | estrogen related receptor beta | ESRRB |
DFNB35, ERR2, ERRb, ESRL2, NR3B2 |
190.5kb | Sense | Chr. 14 - 76371347 - 76501839 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

GPATCH2L
Genomic Map