Assay Details
| Gene Symbol: LINC01284 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110382.1 | 5 - 6 | 272 | 91 | ||
| GenBank mRNA | BX112678.1 | - | 5 - 6 | 275 | 91 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | bone morphogenetic protein 15 | BMP15 |
GDF9B, ODG2, POF4 |
-184.6kb | Anti-sense | Chr. X - 50910735 - 50916641 on NCBI Build GRCh38 |
| Downstream | nudix hydrolase 10 | NUDT10 |
APS2, DIPP3-alpha, DIPP3a |
230.8kb | Anti-sense | Chr. X - 51332231 - 51337525 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

LINC01284
Genomic Map