Assay Details
| Gene Symbol: KANTR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110456.1 | 2 - 3 | 207 | 65 | ||
| GenBank mRNA | BX353482.2 | - | 2 - 3 | 207 | 65 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | TSPY like 2 | TSPYL2 |
CDA1, CINAP, CTCL, DENTT, HRIHFB2216, NP79, SE204, TSPX |
-5.9kb | Sense | Chr. X - 53082327 - 53088546 on NCBI Build GRCh38 |
| Downstream | lysine demethylase 5C | KDM5C |
DXS1272E, JARID1C, MRX13, MRXJ, MRXSCJ, MRXSJ, SMCX, XE169 |
14.7kb | Anti-sense | Chr. X - 53159735 - 53225422 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007311.hg.1 | ENST00000366185.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007311.hg.1 | ENST00000366185.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007311.hg.1 | ENST00000366185.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007311.hg.1 | ENST00000366185.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007311.hg.1 | ENST00000366185.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007311.hg.1 | ENST00000366185.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007311.hg.1 | ENST00000366185.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007311.hg.1 | ENST00000604062.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007311.hg.1 | ENST00000604062.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007311.hg.1 | ENST00000604062.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007311.hg.1 | ENST00000604062.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007311.hg.1 | ENST00000604062.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007311.hg.1 | ENST00000604062.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007311.hg.1 | ENST00000604062.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007311.hg.1 | ENST00000604062.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007311.hg.1 | ENST00000604062.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007311.hg.1 | ENST00000604062.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007311.hg.1 | ENST00000604062.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007311.hg.1 | ENST00000604062.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007311.hg.1 | ENST00000604369.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007311.hg.1 | ENST00000604369.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007311.hg.1 | ENST00000604369.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007311.hg.1 | ENST00000604369.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007311.hg.1 | ENST00000604369.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007311.hg.1 | ENST00000604369.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007311.hg.1 | ENST00000604369.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007311.hg.1 | ENST00000604369.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007311.hg.1 | ENST00000604369.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007311.hg.1 | ENST00000604369.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007311.hg.1 | ENST00000604369.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007311.hg.1 | ENST00000604369.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007311.hg.1 | ENST00000604849.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007311.hg.1 | ENST00000604849.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007311.hg.1 | ENST00000604849.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007311.hg.1 | ENST00000604849.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007311.hg.1 | ENST00000604849.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007311.hg.1 | ENST00000605526.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007311.hg.1 | ENST00000605526.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007311.hg.1 | ENST00000605526.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007311.hg.1 | ENST00000605526.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007311.hg.1 | ENST00000605526.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007311.hg.1 | ENST00000605526.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007311.hg.1 | ENST00000605526.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007311.hg.1 | ENST00000605526.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007311.hg.1 | NR_110456.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007311.hg.1 | NR_110456.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007311.hg.1 | NR_110456.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007311.hg.1 | NR_110456.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007311.hg.1 | NR_110456.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007311.hg.1 | NR_110456.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007311.hg.1 | NR_110456.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007311.hg.1 | NR_110456.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007311.hg.1 | NR_110456.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007311.hg.1 | NR_110456.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007311.hg.1 | NR_110456.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00150357.hg.1 | ENST00000366185.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00150357.hg.1 | ENST00000604849.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00150357.hg.1 | NR_110456.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00150357.hg.1 | ENST00000366185.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00150357.hg.1 | ENST00000604849.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00150357.hg.1 | NR_110456.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00150357.hg.1 | TC0X00007311.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00150353.hg.1 | ENST00000366185.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00150353.hg.1 | ENST00000604849.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00150353.hg.1 | NR_110456.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00150353.hg.1 | ENST00000366185.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00150353.hg.1 | ENST00000604849.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00150353.hg.1 | NR_110456.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00150353.hg.1 | TC0X00007311.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00150354.hg.1 | ENST00000366185.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00150354.hg.1 | NR_110456.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00150354.hg.1 | ENST00000366185.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00150354.hg.1 | NR_110456.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00150354.hg.1 | TC0X00007311.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00150356.hg.1 | ENST00000366185.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00150356.hg.1 | NR_110456.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00150356.hg.1 | ENST00000366185.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00150356.hg.1 | NR_110456.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00150356.hg.1 | TC0X00007311.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00150358.hg.1 | ENST00000604849.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00150358.hg.1 | NR_110456.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00150358.hg.1 | ENST00000604849.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00150358.hg.1 | NR_110456.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00150358.hg.1 | TC0X00007311.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00007311.hg.1 | ENST00000366185.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007311.hg.1 | ENST00000366185.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007311.hg.1 | ENST00000366185.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007311.hg.1 | ENST00000366185.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007311.hg.1 | ENST00000366185.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007311.hg.1 | ENST00000366185.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007311.hg.1 | ENST00000366185.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007311.hg.1 | ENST00000604062.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007311.hg.1 | ENST00000604062.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007311.hg.1 | ENST00000604062.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007311.hg.1 | ENST00000604062.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007311.hg.1 | ENST00000604062.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007311.hg.1 | ENST00000604062.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007311.hg.1 | ENST00000604062.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007311.hg.1 | ENST00000604062.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007311.hg.1 | ENST00000604062.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007311.hg.1 | ENST00000604062.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007311.hg.1 | ENST00000604062.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007311.hg.1 | ENST00000604062.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007311.hg.1 | ENST00000604369.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007311.hg.1 | ENST00000604369.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007311.hg.1 | ENST00000604369.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007311.hg.1 | ENST00000604369.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007311.hg.1 | ENST00000604369.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007311.hg.1 | ENST00000604369.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007311.hg.1 | ENST00000604369.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007311.hg.1 | ENST00000604369.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007311.hg.1 | ENST00000604369.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007311.hg.1 | ENST00000604369.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007311.hg.1 | ENST00000604369.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007311.hg.1 | ENST00000604369.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007311.hg.1 | ENST00000604849.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007311.hg.1 | ENST00000604849.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007311.hg.1 | ENST00000604849.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007311.hg.1 | ENST00000604849.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007311.hg.1 | ENST00000604849.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007311.hg.1 | ENST00000605526.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007311.hg.1 | ENST00000605526.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007311.hg.1 | ENST00000605526.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007311.hg.1 | ENST00000605526.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007311.hg.1 | ENST00000605526.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007311.hg.1 | ENST00000605526.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007311.hg.1 | ENST00000605526.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007311.hg.1 | ENST00000605526.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007311.hg.1 | NR_110456.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007311.hg.1 | NR_110456.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007311.hg.1 | NR_110456.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007311.hg.1 | NR_110456.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007311.hg.1 | NR_110456.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007311.hg.1 | NR_110456.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007311.hg.1 | NR_110456.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007311.hg.1 | NR_110456.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007311.hg.1 | NR_110456.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007311.hg.1 | NR_110456.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007311.hg.1 | NR_110456.2 | Refseq |

KANTR
Genomic Map