Assay Details
| Gene Symbol: LINC01500 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110547.1 | 1 - 2 | 274 | 73 | ||
| GenBank mRNA | CR739775.1 | - | 1 - 2 | 274 | 73 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | dishevelled binding antagonist of beta catenin 1 | DACT1 |
DAPPER, DAPPER1, DPR1, FRODO, HDPR1, THYEX3 |
-180.0kb | Sense | Chr. 14 - 58634068 - 58648321 on NCBI Build GRCh38 |
| Downstream | dishevelled associated activator of morphogenesis 1 | DAAM1 | 294.3kb | Sense | Chr. 14 - 59188611 - 59371405 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000553762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000553762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000553762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000553762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000553762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000553762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000555378.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000555378.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000555378.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000555378.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000555378.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000555378.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000555378.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000555378.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000555378.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000555378.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000555378.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000555378.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000556026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000556026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000556026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000556026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000556026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000556815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000556815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000556815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000556815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000556815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000556815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000556815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000556815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000556815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000556815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000556815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000556815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000556815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000556815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | NR_110547.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | NR_110547.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | NR_110547.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | NR_110547.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | NR_110547.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | NR_110547.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | NR_110547.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000553762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000553762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000553762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000553762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000553762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000553762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000555378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000555378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000555378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000555378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000555378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000555378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000555378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000555378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000555378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000555378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000555378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000555378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000556026.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000556026.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000556026.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000556026.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000556026.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000556815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000556815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000556815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000556815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000556815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000556815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000556815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000556815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000556815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000556815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000556815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000556815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000556815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000556815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | NR_110547.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | NR_110547.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | NR_110547.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | NR_110547.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | NR_110547.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | NR_110547.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | NR_110547.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400150842.hg.1 | ENST00000553762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150842.hg.1 | ENST00000555378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150842.hg.1 | ENST00000555378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150842.hg.1 | ENST00000556815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150842.hg.1 | ENST00000556815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150842.hg.1 | NR_110547.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400150842.hg.1 | ENST00000553762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150842.hg.1 | ENST00000555378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150842.hg.1 | ENST00000555378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150842.hg.1 | ENST00000556815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150842.hg.1 | ENST00000556815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150842.hg.1 | NR_110547.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400150842.hg.1 | TC1400007323.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400150846.hg.1 | ENST00000553762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150846.hg.1 | ENST00000555378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150846.hg.1 | ENST00000555378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150846.hg.1 | ENST00000556026.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150846.hg.1 | ENST00000556815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150846.hg.1 | ENST00000556815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150846.hg.1 | NR_110547.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400150846.hg.1 | ENST00000553762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150846.hg.1 | ENST00000555378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150846.hg.1 | ENST00000555378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150846.hg.1 | ENST00000556026.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150846.hg.1 | ENST00000556815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150846.hg.1 | ENST00000556815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150846.hg.1 | NR_110547.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400150846.hg.1 | TC1400007323.hg.1 | Clariom D TC |

LINC01500
Genomic Map