Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008323.hg.1 |
ENST00000504004.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008323.hg.1 |
ENST00000504004.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008323.hg.1 |
ENST00000504004.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008323.hg.1 |
ENST00000504004.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008323.hg.1 |
NR_110559.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008323.hg.1 |
NR_110559.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008323.hg.1 |
NR_110559.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008323.hg.1 |
NR_110559.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500156960.hg.1 |
ENST00000504004.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500156960.hg.1 |
NR_110559.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500156960.hg.1 |
ENST00000504004.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500156960.hg.1 |
NR_110559.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500156960.hg.1 |
TC0500008323.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500008323.hg.1 |
ENST00000504004.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008323.hg.1 |
ENST00000504004.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008323.hg.1 |
ENST00000504004.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008323.hg.1 |
ENST00000504004.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008323.hg.1 |
NR_110559.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008323.hg.1 |
NR_110559.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008323.hg.1 |
NR_110559.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008323.hg.1 |
NR_110559.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500156961.hg.1 |
ENST00000504004.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500156961.hg.1 |
NR_110559.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500156961.hg.1 |
ENST00000504004.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500156961.hg.1 |
NR_110559.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500156961.hg.1 |
TC0500008323.hg.1 |
Clariom D TC |