Assay Details
| Gene Symbol: LANCL1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110604.1 | 5 - 6 | 516 | 87 | ||
| NR_110605.1 | 5 - 6 | 489 | 87 | |||
| NR_110606.1 | 4 - 5 | 331 | 87 | |||
| GenBank mRNA | AL599033.1 | - | 5 - 6 | 459 | 87 | |
| DA820426.1 | - | 4 - 5 | 331 | 87 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | LanC like 1 | LANCL1 |
GPR69A, p40 |
Anti-sense | Chr. 2 - 210431249 - 210477618 on NCBI Build GRCh38 | |
| Downstream | carbamoyl-phosphate synthase 1 | CPS1 |
CPSASE1, PHN |
7.3kb | Sense | Chr. 2 - 210477682 - 210679107 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LANCL1-AS1
Genomic Map