| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006704.hg.1 |
ENST00000417299.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006704.hg.1 |
ENST00000417299.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006704.hg.1 |
NR_110611.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006704.hg.1 |
NR_110611.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006704.hg.1 |
NR_110611.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000145959.hg.1 |
NR_110611.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000145959.hg.1 |
TC2000006704.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000145959.hg.1 |
NR_110611.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000145958.hg.1 |
ENST00000417299.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000145958.hg.1 |
NR_110611.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000145958.hg.1 |
TC2000006704.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000145958.hg.1 |
ENST00000417299.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000145958.hg.1 |
NR_110611.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000145956.hg.1 |
ENST00000417299.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000145956.hg.1 |
NR_110611.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000145956.hg.1 |
TC2000006704.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000145956.hg.1 |
ENST00000417299.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000145956.hg.1 |
NR_110611.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006704.hg.1 |
ENST00000417299.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006704.hg.1 |
ENST00000417299.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006704.hg.1 |
NR_110611.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006704.hg.1 |
NR_110611.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006704.hg.1 |
NR_110611.1 |
Refseq |