Assay Details
Gene Symbol: NFIA-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110618.1 | 2 - 3 | 366 | 125 | ||
GenBank mRNA | DA722202.1 | - | 2 - 3 | 233 | 125 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC101926964 | LOC101926964 | -128.9kb | Sense | Chr. 1 - 60659631 - 60825584 on NCBI Build GRCh38 | |
Downstream | nuclear factor I A | NFIA |
CTF, NF-I/A, NF1-A, NFI-A, NFI-L |
107.2kb | Anti-sense | Chr. 1 - 61077124 - 61462788 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() |
