Assay Details
| Gene Symbol: ALS2CR12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110620.1 | 6 - 7 | 1079 | 96 | ||
| XM_011510612.2 | XP_011508914.1 | 1 - 2 | 85 | 96 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | caspase 8 | CASP8 |
ALPS2B, CAP4, Casp-8, FLICE, MACH, MCH5 |
-43.0kb | Anti-sense | Chr. 2 - 201233443 - 201287711 on NCBI Build GRCh38 |
| Downstream | trafficking kinesin protein 2 | TRAK2 |
ALS2CR3, CALS-C, GRIF-1, GRIF1, MILT2, OIP98 |
32.9kb | Sense | Chr. 2 - 201377207 - 201451596 on NCBI Build GRCh38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011510612.2 transcripts.
Set Membership: |
|

ALS2CR12
Genomic Map