| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009121.hg.1 |
ENST00000411798.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009121.hg.1 |
ENST00000411798.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009121.hg.1 |
ENST00000411798.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009121.hg.1 |
ENST00000440116.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009121.hg.1 |
ENST00000440116.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009121.hg.1 |
ENST00000440116.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009121.hg.1 |
ENST00000456933.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009121.hg.1 |
ENST00000456933.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009121.hg.1 |
ENST00000456933.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009121.hg.1 |
ENST00000456933.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009121.hg.1 |
NR_110621.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009121.hg.1 |
NR_110621.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009121.hg.1 |
NR_110621.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009121.hg.1 |
NR_110621.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009121.hg.1 |
NR_110621.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009121.hg.1 |
NR_110622.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009121.hg.1 |
NR_110622.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009121.hg.1 |
NR_110622.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009121.hg.1 |
NR_110622.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009121.hg.1 |
ENST00000411798.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009121.hg.1 |
ENST00000411798.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009121.hg.1 |
ENST00000411798.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009121.hg.1 |
ENST00000440116.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009121.hg.1 |
ENST00000440116.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009121.hg.1 |
ENST00000440116.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009121.hg.1 |
ENST00000456933.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009121.hg.1 |
ENST00000456933.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009121.hg.1 |
ENST00000456933.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009121.hg.1 |
ENST00000456933.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009121.hg.1 |
NR_110621.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009121.hg.1 |
NR_110621.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009121.hg.1 |
NR_110621.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009121.hg.1 |
NR_110621.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009121.hg.1 |
NR_110621.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009121.hg.1 |
NR_110622.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009121.hg.1 |
NR_110622.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009121.hg.1 |
NR_110622.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009121.hg.1 |
NR_110622.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100165093.hg.1 |
ENST00000411798.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100165093.hg.1 |
NR_110621.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100165093.hg.1 |
ENST00000411798.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100165093.hg.1 |
NR_110621.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100165093.hg.1 |
TC0100009121.hg.1 |
Clariom D TC |