| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014735.hg.1 |
ENST00000421931.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014735.hg.1 |
ENST00000421931.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014735.hg.1 |
ENST00000421931.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014735.hg.1 |
ENST00000421931.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014735.hg.1 |
ENST00000421931.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014735.hg.1 |
ENST00000452834.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014735.hg.1 |
ENST00000452834.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014735.hg.1 |
ENST00000452834.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014735.hg.1 |
ENST00000452834.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014735.hg.1 |
ENST00000452834.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014735.hg.1 |
ENST00000452834.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014735.hg.1 |
ENST00000452834.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014735.hg.1 |
ENST00000452834.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014735.hg.1 |
ENST00000452834.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014735.hg.1 |
ENST00000452834.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014735.hg.1 |
ENST00000452834.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014735.hg.1 |
ENST00000452834.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014735.hg.1 |
NR_110632.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014735.hg.1 |
NR_110632.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014735.hg.1 |
NR_110632.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014735.hg.1 |
NR_110632.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014735.hg.1 |
NR_110632.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014735.hg.1 |
NR_110632.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014735.hg.1 |
NR_110633.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014735.hg.1 |
NR_110633.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014735.hg.1 |
NR_110633.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014735.hg.1 |
NR_110633.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014735.hg.1 |
NR_110633.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014735.hg.1 |
NR_110633.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014735.hg.1 |
ENST00000421931.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014735.hg.1 |
ENST00000421931.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014735.hg.1 |
ENST00000421931.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014735.hg.1 |
ENST00000421931.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014735.hg.1 |
ENST00000421931.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014735.hg.1 |
ENST00000452834.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014735.hg.1 |
ENST00000452834.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014735.hg.1 |
ENST00000452834.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014735.hg.1 |
ENST00000452834.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014735.hg.1 |
ENST00000452834.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014735.hg.1 |
ENST00000452834.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014735.hg.1 |
ENST00000452834.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014735.hg.1 |
ENST00000452834.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014735.hg.1 |
ENST00000452834.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014735.hg.1 |
ENST00000452834.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014735.hg.1 |
ENST00000452834.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014735.hg.1 |
ENST00000452834.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014735.hg.1 |
NR_110632.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014735.hg.1 |
NR_110632.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014735.hg.1 |
NR_110632.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014735.hg.1 |
NR_110632.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014735.hg.1 |
NR_110632.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014735.hg.1 |
NR_110632.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014735.hg.1 |
NR_110633.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014735.hg.1 |
NR_110633.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014735.hg.1 |
NR_110633.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014735.hg.1 |
NR_110633.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014735.hg.1 |
NR_110633.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014735.hg.1 |
NR_110633.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100209639.hg.1 |
ENST00000421931.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100209639.hg.1 |
NR_110633.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100209639.hg.1 |
ENST00000421931.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100209639.hg.1 |
NR_110633.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100209639.hg.1 |
TC0100014735.hg.1 |
Clariom D TC |