Assay Details
| Gene Symbol: LINC01365 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110660.1 | 4 - 5 | 407 | 149 | ||
| GenBank mRNA | BC038750.1 | - | 4 - 5 | 407 | 149 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | phosphodiesterase 5A | PDE5A |
CGB-PDE, CN5A, PDE5 |
-170.3kb | Sense | Chr. 4 - 119494395 - 119629080 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC100996694 | LOC100996694 | 138.8kb | Anti-sense | Chr. 4 - 119939540 - 119964293 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LINC01365
Genomic Map