| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010023.hg.1 |
ENST00000427145.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010023.hg.1 |
ENST00000427145.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010023.hg.1 |
ENST00000427145.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010023.hg.1 |
NR_110685.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010023.hg.1 |
NR_110685.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010023.hg.1 |
NR_110685.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010023.hg.1 |
ENST00000427145.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010023.hg.1 |
ENST00000427145.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010023.hg.1 |
ENST00000427145.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010023.hg.1 |
NR_110685.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010023.hg.1 |
NR_110685.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010023.hg.1 |
NR_110685.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100172642.hg.1 |
ENST00000427145.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100172642.hg.1 |
NR_110685.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100172642.hg.1 |
ENST00000427145.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100172642.hg.1 |
NR_110685.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100172642.hg.1 |
TC0100010023.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100172641.hg.1 |
ENST00000427145.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100172641.hg.1 |
NR_110685.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100172641.hg.1 |
ENST00000427145.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100172641.hg.1 |
NR_110685.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100172641.hg.1 |
TC0100010023.hg.1 |
Clariom D TC |