Assay Details
| Gene Symbol: HIPK1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110726.1 | 2 - 3 | 305 | 106 | ||
| GenBank mRNA | BC048113.1 | - | 1 - 2 | 985 | 106 | |
| HY108301.1 | - | 2 - 3 | 305 | 106 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | DNA cross-link repair 1B | DCLRE1B |
APOLLO, SNM1B, SNMIB |
-9.9kb | Anti-sense | Chr. 1 - 113904619 - 113914086 on NCBI Build GRCh38 |
| Downstream | homeodomain interacting protein kinase 1 | HIPK1 |
Myak, Nbak2 |
809.0 | Anti-sense | Chr. 1 - 113929197 - 113977869 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

HIPK1-AS1
Genomic Map