Assay Details
| Gene Symbol: LINC01543 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110762.1 | 3 - 4 | 616 | 85 | ||
| GenBank mRNA | HY185322.1 | - | 1 - 2 | 85 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | TATA-box binding protein associated factor 4b | TAF4B |
SPGF13, TAF2C2, TAFII105 |
-34.2kb | Sense | Chr. 18 - 26226491 - 26391685 on NCBI Build GRCh38 |
| Downstream | potassium channel tetramerization domain containing 1 | KCTD1 |
C18orf5, SENS |
16.9kb | Anti-sense | Chr. 18 - 26454910 - 26657401 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

LINC01543
Genomic Map