| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006645.hg.1 |
ENST00000582435.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006645.hg.1 |
ENST00000582435.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006645.hg.1 |
ENST00000582435.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006645.hg.1 |
NR_110773.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006645.hg.1 |
NR_110773.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006645.hg.1 |
NR_110773.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006645.hg.1 |
NR_110774.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006645.hg.1 |
NR_110774.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006645.hg.1 |
NR_110774.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800006645.hg.1 |
ENST00000582435.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800006645.hg.1 |
ENST00000582435.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800006645.hg.1 |
ENST00000582435.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800006645.hg.1 |
NR_110773.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800006645.hg.1 |
NR_110773.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800006645.hg.1 |
NR_110773.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800006645.hg.1 |
NR_110774.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800006645.hg.1 |
NR_110774.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800006645.hg.1 |
NR_110774.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800145441.hg.1 |
ENST00000582435.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800145441.hg.1 |
NR_110773.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800145441.hg.1 |
NR_110774.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800145441.hg.1 |
ENST00000582435.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800145441.hg.1 |
NR_110773.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800145441.hg.1 |
NR_110774.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800145441.hg.1 |
TC1800006645.hg.1 |
Clariom D TC |