Assay Details
| Gene Symbol: LOC102724651 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110798.1 | 3 - 4 | 272 | 109 | ||
| GenBank mRNA | BX088865.1 | - | 3 - 4 | 236 | 109 | |
| DB079133.1 | - | 3 - 4 | 273 | 109 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101928167 | LOC101928167 | -1.5kb | Anti-sense | Chr. 18 - 53579623 - 53581059 on NCBI Build GRCh38 | |
| Downstream | methyl-CpG binding domain protein 2 | MBD2 |
DMTase, NY-CO-41 |
567.3kb | Anti-sense | Chr. 18 - 54151601 - 54224788 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC102724651
Genomic Map