Assay Details
| Gene Symbol: LOC101928163 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110799.1 | 2 - 3 | 194 | 131 | ||
| GenBank mRNA | AK310251.1 | - | 2 - 3 | 194 | 131 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
ArfGAP with SH3 domain, ankyrin repeat and PH domain 3 |
ASAP3 |
ACAP4, CENTB6, DDEFL1, UPLC1 |
-42.4kb | Anti-sense | Chr. 1 - 23428563 - 23484631 on NCBI Build GRCh38 |
| Downstream |
inhibitor of DNA binding 3, HLH protein |
ID3 |
HEIR-1, bHLHb25 |
29.5kb | Anti-sense | Chr. 1 - 23557930 - 23559794 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

LOC101928163
Genomic Map