Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011225.hg.1 |
ENST00000578662.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011225.hg.1 |
ENST00000578662.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011225.hg.1 |
ENST00000578662.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011225.hg.1 |
NR_110807.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011225.hg.1 |
NR_110807.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011225.hg.1 |
NR_110807.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700190150.hg.1 |
ENST00000578662.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700190150.hg.1 |
NR_110807.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700190150.hg.1 |
ENST00000578662.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700190150.hg.1 |
NR_110807.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700190150.hg.1 |
TC1700011225.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700011225.hg.1 |
ENST00000578662.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011225.hg.1 |
ENST00000578662.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011225.hg.1 |
ENST00000578662.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011225.hg.1 |
NR_110807.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011225.hg.1 |
NR_110807.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011225.hg.1 |
NR_110807.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700190149.hg.1 |
ENST00000578662.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700190149.hg.1 |
NR_110807.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700190149.hg.1 |
ENST00000578662.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700190149.hg.1 |
NR_110807.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700190149.hg.1 |
TC1700011225.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700190151.hg.1 |
ENST00000578662.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700190151.hg.1 |
NR_110807.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700190151.hg.1 |
ENST00000578662.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700190151.hg.1 |
NR_110807.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700190151.hg.1 |
TC1700011225.hg.1 |
Clariom D TC |