| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012288.hg.1 |
ENST00000586209.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012288.hg.1 |
ENST00000586209.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012288.hg.1 |
ENST00000586209.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012288.hg.1 |
ENST00000586209.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012288.hg.1 |
ENST00000593015.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012288.hg.1 |
ENST00000593015.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012288.hg.1 |
ENST00000593015.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012288.hg.1 |
ENST00000593015.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012288.hg.1 |
ENST00000593015.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012288.hg.1 |
ENST00000593015.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012288.hg.1 |
ENST00000593015.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012288.hg.1 |
ENST00000593015.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012288.hg.1 |
NR_110815.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012288.hg.1 |
NR_110815.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012288.hg.1 |
NR_110815.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012288.hg.1 |
NR_110815.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012288.hg.1 |
NR_110816.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012288.hg.1 |
NR_110816.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012288.hg.1 |
NR_110816.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700203378.hg.1 |
ENST00000586209.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700203378.hg.1 |
NR_110815.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700203378.hg.1 |
ENST00000586209.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700203378.hg.1 |
NR_110815.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700203378.hg.1 |
TC1700012288.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1700012288.hg.1 |
ENST00000586209.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012288.hg.1 |
ENST00000586209.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012288.hg.1 |
ENST00000586209.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012288.hg.1 |
ENST00000586209.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012288.hg.1 |
ENST00000593015.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012288.hg.1 |
ENST00000593015.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012288.hg.1 |
ENST00000593015.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012288.hg.1 |
ENST00000593015.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012288.hg.1 |
ENST00000593015.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012288.hg.1 |
ENST00000593015.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012288.hg.1 |
ENST00000593015.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012288.hg.1 |
ENST00000593015.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012288.hg.1 |
NR_110815.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700012288.hg.1 |
NR_110815.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700012288.hg.1 |
NR_110815.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700012288.hg.1 |
NR_110815.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700012288.hg.1 |
NR_110816.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700012288.hg.1 |
NR_110816.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700012288.hg.1 |
NR_110816.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700203374.hg.1 |
ENST00000586209.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700203374.hg.1 |
ENST00000593015.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700203374.hg.1 |
ENST00000593015.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700203374.hg.1 |
NR_110815.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700203374.hg.1 |
NR_110816.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700203374.hg.1 |
ENST00000586209.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700203374.hg.1 |
ENST00000593015.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700203374.hg.1 |
ENST00000593015.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700203374.hg.1 |
NR_110815.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700203374.hg.1 |
NR_110816.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700203374.hg.1 |
TC1700012288.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700203376.hg.1 |
ENST00000593015.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700203376.hg.1 |
ENST00000593015.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700203376.hg.1 |
NR_110816.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700203376.hg.1 |
ENST00000593015.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700203376.hg.1 |
ENST00000593015.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700203376.hg.1 |
NR_110816.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700203376.hg.1 |
TC1700012288.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700203375.hg.1 |
ENST00000586209.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700203375.hg.1 |
ENST00000593015.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700203375.hg.1 |
ENST00000593015.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700203375.hg.1 |
NR_110815.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700203375.hg.1 |
NR_110816.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700203375.hg.1 |
ENST00000586209.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700203375.hg.1 |
ENST00000593015.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700203375.hg.1 |
ENST00000593015.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700203375.hg.1 |
NR_110815.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700203375.hg.1 |
NR_110816.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700203375.hg.1 |
TC1700012288.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700203379.hg.1 |
ENST00000586209.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700203379.hg.1 |
NR_110815.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700203379.hg.1 |
ENST00000586209.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700203379.hg.1 |
NR_110815.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700203379.hg.1 |
TC1700012288.hg.1 |
Clariom D TC |