Assay Details
| Gene Symbol: PRKCA-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110822.1 | 1 - 2 | 313 | 69 | ||
| GenBank mRNA | CR745158.1 | - | 1 - 2 | 250 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700011498.hg.1 | ENST00000583421.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011498.hg.1 | ENST00000583421.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011498.hg.1 | ENST00000584614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011498.hg.1 | ENST00000584614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011498.hg.1 | ENST00000584715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011498.hg.1 | ENST00000584715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011498.hg.1 | ENST00000584715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011498.hg.1 | ENST00000584715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011498.hg.1 | ENST00000584715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011498.hg.1 | ENST00000584715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011498.hg.1 | ENST00000584715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011498.hg.1 | ENST00000584715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011498.hg.1 | ENST00000584715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011498.hg.1 | ENST00000584715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011498.hg.1 | ENST00000584715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011498.hg.1 | ENST00000584715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011498.hg.1 | ENST00000584715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011498.hg.1 | ENST00000584715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011498.hg.1 | NR_110822.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011498.hg.1 | NR_110822.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011498.hg.1 | NR_110822.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011498.hg.1 | NR_110822.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011498.hg.1 | NR_110822.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011498.hg.1 | NR_110822.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700193152.hg.1 | ENST00000583421.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700193152.hg.1 | ENST00000584715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700193152.hg.1 | ENST00000584715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700193152.hg.1 | NR_110822.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700193152.hg.1 | ENST00000583421.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700193152.hg.1 | ENST00000584715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700193152.hg.1 | ENST00000584715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700193152.hg.1 | NR_110822.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700193152.hg.1 | TC1700011498.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1700011498.hg.1 | ENST00000583421.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011498.hg.1 | ENST00000583421.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011498.hg.1 | ENST00000584614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011498.hg.1 | ENST00000584614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011498.hg.1 | ENST00000584715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011498.hg.1 | ENST00000584715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011498.hg.1 | ENST00000584715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011498.hg.1 | ENST00000584715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011498.hg.1 | ENST00000584715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011498.hg.1 | ENST00000584715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011498.hg.1 | ENST00000584715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011498.hg.1 | ENST00000584715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011498.hg.1 | ENST00000584715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011498.hg.1 | ENST00000584715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011498.hg.1 | ENST00000584715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011498.hg.1 | ENST00000584715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011498.hg.1 | ENST00000584715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011498.hg.1 | ENST00000584715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011498.hg.1 | NR_110822.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011498.hg.1 | NR_110822.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011498.hg.1 | NR_110822.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011498.hg.1 | NR_110822.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011498.hg.1 | NR_110822.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011498.hg.1 | NR_110822.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700193151.hg.1 | ENST00000584715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700193151.hg.1 | ENST00000584715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700193151.hg.1 | NR_110822.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700193151.hg.1 | ENST00000584715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700193151.hg.1 | ENST00000584715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700193151.hg.1 | NR_110822.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700193151.hg.1 | TC1700011498.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700193154.hg.1 | ENST00000583421.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700193154.hg.1 | ENST00000584715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700193154.hg.1 | ENST00000584715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700193154.hg.1 | NR_110822.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700193154.hg.1 | ENST00000583421.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700193154.hg.1 | ENST00000584715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700193154.hg.1 | ENST00000584715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700193154.hg.1 | NR_110822.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700193154.hg.1 | TC1700011498.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700193155.hg.1 | ENST00000584715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700193155.hg.1 | ENST00000584715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700193155.hg.1 | NR_110822.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700193155.hg.1 | ENST00000584715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700193155.hg.1 | ENST00000584715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700193155.hg.1 | NR_110822.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700193155.hg.1 | TC1700011498.hg.1 | Clariom D TC |

PRKCA-AS1
Genomic Map