Assay Details
| Gene Symbol: TSPAN10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290212.1 | NP_001277141.1 | 2 - 3 | 381 | 100 | |
| NM_031945.4 | NP_114151.3 | 1 - 2 | 147 | 100 | ||
| NR_110866.1 | 1 - 2 | 147 | 100 | |||
| GenBank mRNA | AB593081.1 | - | 2 - 3 | 354 | 100 | |
| AB593082.1 | - | 1 - 2 | 142 | 100 | ||
| AF325213.1 | - | 1 - 2 | 103 | 100 | ||
| AK096926.1 | - | 1 - 2 | 129 | 100 | ||
| BC021923.1 | - | 1 - 2 | 116 | 4876887 | 100 | |
| BC032802.1 | - | 1 - 2 | 118 | 5191718 | 100 | |
| BG760374.1 | - | 1 - 2 | 116 | 100 | ||
| DA912496.1 | - | 1 - 2 | 129 | 100 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AB593082.1, AF325213.1, BC021923.1, BC032802.1, NM_031945.4 transcripts.
Set Membership: |
|

TSPAN10
Genomic Map