Assay Details
| Gene Symbol: LOC101928936 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110867.1 | 1 - 2 | 184 | 95 | ||
| GenBank mRNA | AK131305.1 | - | 1 - 2 | 184 | 95 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cannabinoid receptor 1 | CNR1 |
CANN6, CB-R, CB1, CB1A, CB1K5, CB1R, CNR |
-363.2kb | Sense | Chr. 6 - 88139864 - 88167429 on NCBI Build GRCh38 |
| Downstream | RNA guanylyltransferase and 5'-phosphatase | RNGTT |
CAP1A, HCE, HCE1, hCAP |
70.5kb | Sense | Chr. 6 - 88609897 - 88963716 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

LOC101928936
Genomic Map