| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011431.hg.1 |
ENST00000512279.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011431.hg.1 |
ENST00000512279.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011431.hg.1 |
ENST00000512279.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011431.hg.1 |
ENST00000512279.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011431.hg.1 |
ENST00000512279.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011431.hg.1 |
ENST00000512279.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011431.hg.1 |
NR_110934.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011431.hg.1 |
NR_110934.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011431.hg.1 |
NR_110934.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011431.hg.1 |
NR_110934.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011431.hg.1 |
NR_110934.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011431.hg.1 |
NR_110934.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600191513.hg.1 |
ENST00000512279.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600191513.hg.1 |
NR_110934.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600191513.hg.1 |
ENST00000512279.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600191513.hg.1 |
NR_110934.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600191513.hg.1 |
TC1600011431.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1600011431.hg.1 |
ENST00000512279.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011431.hg.1 |
ENST00000512279.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011431.hg.1 |
ENST00000512279.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011431.hg.1 |
ENST00000512279.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011431.hg.1 |
ENST00000512279.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011431.hg.1 |
ENST00000512279.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011431.hg.1 |
NR_110934.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011431.hg.1 |
NR_110934.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011431.hg.1 |
NR_110934.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011431.hg.1 |
NR_110934.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011431.hg.1 |
NR_110934.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011431.hg.1 |
NR_110934.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600191509.hg.1 |
ENST00000512279.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600191509.hg.1 |
NR_110934.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600191509.hg.1 |
ENST00000512279.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600191509.hg.1 |
NR_110934.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600191509.hg.1 |
TC1600011431.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600191512.hg.1 |
ENST00000512279.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600191512.hg.1 |
NR_110934.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600191512.hg.1 |
ENST00000512279.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600191512.hg.1 |
NR_110934.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600191512.hg.1 |
TC1600011431.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600191510.hg.1 |
ENST00000512279.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600191510.hg.1 |
NR_110934.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600191510.hg.1 |
ENST00000512279.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600191510.hg.1 |
NR_110934.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600191510.hg.1 |
TC1600011431.hg.1 |
Clariom D TC |