Assay Details
Gene Symbol: LINC01504 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110952.1 | 1 - 2 | 192 | 75 | ||
NR_110953.1 | 1 - 2 | 192 | 75 | |||
GenBank mRNA | AA436218.1 | - | 1 - 2 | 110 | 75 | |
AK095210.1 | - | 1 - 2 | 192 | 75 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | guanine deaminase | GDA |
CYPIN, GUANASE, NEDASIN |
-46.4kb | Sense | Chr. 9 - 72114595 - 72256045 on NCBI Build GRCh38 |
Downstream | zinc finger AN1-type containing 5 | ZFAND5 |
ZA20D2, ZFAND5A, ZNF216 |
36.3kb | Anti-sense | Chr. 9 - 72351425 - 72365247 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0900007556.hg.1 | ENST00000436054.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007556.hg.1 | ENST00000436054.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007556.hg.1 | ENST00000436054.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007556.hg.1 | ENST00000436054.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007556.hg.1 | ENST00000436054.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007556.hg.1 | ENST00000436054.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007556.hg.1 | ENST00000436054.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007556.hg.1 | ENST00000436054.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007556.hg.1 | ENST00000436054.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007556.hg.1 | ENST00000436054.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007556.hg.1 | ENST00000436054.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007556.hg.1 | ENST00000451152.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007556.hg.1 | ENST00000451152.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007556.hg.1 | ENST00000451152.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007556.hg.1 | ENST00000451152.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007556.hg.1 | ENST00000451152.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007556.hg.1 | ENST00000451152.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007556.hg.1 | ENST00000451152.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007556.hg.1 | ENST00000451596.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007556.hg.1 | ENST00000451596.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007556.hg.1 | ENST00000451596.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007556.hg.1 | ENST00000451596.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007556.hg.1 | ENST00000451596.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007556.hg.1 | ENST00000451596.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007556.hg.1 | ENST00000451596.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007556.hg.1 | ENST00000451596.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007556.hg.1 | NR_110952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007556.hg.1 | NR_110952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007556.hg.1 | NR_110952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007556.hg.1 | NR_110952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007556.hg.1 | NR_110952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007556.hg.1 | NR_110952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007556.hg.1 | NR_110952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007556.hg.1 | NR_110952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007556.hg.1 | NR_110952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007556.hg.1 | NR_110952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007556.hg.1 | NR_110952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007556.hg.1 | NR_110953.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007556.hg.1 | NR_110953.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007556.hg.1 | NR_110953.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007556.hg.1 | NR_110953.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007556.hg.1 | NR_110953.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007556.hg.1 | NR_110953.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007556.hg.1 | NR_110953.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007556.hg.1 | NR_110953.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900151138.hg.1 | ENST00000436054.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900151138.hg.1 | ENST00000451152.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900151138.hg.1 | ENST00000451596.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900151138.hg.1 | NR_110952.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900151138.hg.1 | NR_110953.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900151138.hg.1 | ENST00000436054.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900151138.hg.1 | ENST00000451152.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900151138.hg.1 | ENST00000451596.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900151138.hg.1 | NR_110952.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900151138.hg.1 | NR_110953.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900151138.hg.1 | TC0900007556.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900151139.hg.1 | ENST00000436054.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900151139.hg.1 | ENST00000451152.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900151139.hg.1 | ENST00000451596.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900151139.hg.1 | NR_110952.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900151139.hg.1 | NR_110953.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900151139.hg.1 | ENST00000436054.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900151139.hg.1 | ENST00000451152.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900151139.hg.1 | ENST00000451596.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900151139.hg.1 | NR_110952.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900151139.hg.1 | NR_110953.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900151139.hg.1 | TC0900007556.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900151141.hg.1 | ENST00000451152.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900151141.hg.1 | ENST00000451596.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900151141.hg.1 | NR_110953.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900151141.hg.1 | ENST00000451152.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900151141.hg.1 | ENST00000451596.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900151141.hg.1 | NR_110953.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900151141.hg.1 | TC0900007556.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0900007556.hg.1 | ENST00000436054.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007556.hg.1 | ENST00000436054.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007556.hg.1 | ENST00000436054.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007556.hg.1 | ENST00000436054.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007556.hg.1 | ENST00000436054.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007556.hg.1 | ENST00000436054.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007556.hg.1 | ENST00000436054.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007556.hg.1 | ENST00000436054.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007556.hg.1 | ENST00000436054.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007556.hg.1 | ENST00000436054.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007556.hg.1 | ENST00000436054.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007556.hg.1 | ENST00000451152.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007556.hg.1 | ENST00000451152.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007556.hg.1 | ENST00000451152.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007556.hg.1 | ENST00000451152.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007556.hg.1 | ENST00000451152.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007556.hg.1 | ENST00000451152.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007556.hg.1 | ENST00000451152.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007556.hg.1 | ENST00000451596.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007556.hg.1 | ENST00000451596.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007556.hg.1 | ENST00000451596.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007556.hg.1 | ENST00000451596.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007556.hg.1 | ENST00000451596.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007556.hg.1 | ENST00000451596.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007556.hg.1 | ENST00000451596.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007556.hg.1 | ENST00000451596.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007556.hg.1 | NR_110952.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007556.hg.1 | NR_110952.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007556.hg.1 | NR_110952.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007556.hg.1 | NR_110952.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007556.hg.1 | NR_110952.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007556.hg.1 | NR_110952.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007556.hg.1 | NR_110952.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007556.hg.1 | NR_110952.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007556.hg.1 | NR_110952.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007556.hg.1 | NR_110952.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007556.hg.1 | NR_110952.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007556.hg.1 | NR_110953.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007556.hg.1 | NR_110953.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007556.hg.1 | NR_110953.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007556.hg.1 | NR_110953.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007556.hg.1 | NR_110953.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007556.hg.1 | NR_110953.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007556.hg.1 | NR_110953.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007556.hg.1 | NR_110953.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900151146.hg.1 | ENST00000436054.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900151146.hg.1 | NR_110952.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900151146.hg.1 | ENST00000436054.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900151146.hg.1 | NR_110952.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900151146.hg.1 | TC0900007556.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900151153.hg.1 | ENST00000436054.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900151153.hg.1 | NR_110952.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900151153.hg.1 | ENST00000436054.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900151153.hg.1 | NR_110952.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900151153.hg.1 | TC0900007556.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900151133.hg.1 | ENST00000436054.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900151133.hg.1 | ENST00000451152.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900151133.hg.1 | ENST00000451596.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900151133.hg.1 | NR_110952.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900151133.hg.1 | NR_110953.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900151133.hg.1 | ENST00000436054.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900151133.hg.1 | ENST00000451152.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900151133.hg.1 | ENST00000451596.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900151133.hg.1 | NR_110952.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900151133.hg.1 | NR_110953.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900151133.hg.1 | TC0900007556.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900151148.hg.1 | ENST00000436054.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900151148.hg.1 | NR_110952.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900151148.hg.1 | ENST00000436054.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900151148.hg.1 | NR_110952.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900151148.hg.1 | TC0900007556.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900151150.hg.1 | ENST00000436054.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900151150.hg.1 | NR_110952.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900151150.hg.1 | ENST00000436054.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900151150.hg.1 | NR_110952.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900151150.hg.1 | TC0900007556.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900151130.hg.1 | ENST00000436054.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900151130.hg.1 | ENST00000451596.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900151130.hg.1 | NR_110952.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900151130.hg.1 | NR_110953.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900151130.hg.1 | ENST00000436054.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900151130.hg.1 | ENST00000451596.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900151130.hg.1 | NR_110952.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900151130.hg.1 | NR_110953.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900151130.hg.1 | TC0900007556.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900151142.hg.1 | ENST00000451152.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900151142.hg.1 | ENST00000451596.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900151142.hg.1 | NR_110953.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900151142.hg.1 | ENST00000451152.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900151142.hg.1 | ENST00000451596.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900151142.hg.1 | NR_110953.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900151142.hg.1 | TC0900007556.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900151152.hg.1 | ENST00000436054.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900151152.hg.1 | NR_110952.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900151152.hg.1 | ENST00000436054.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900151152.hg.1 | NR_110952.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900151152.hg.1 | TC0900007556.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900151140.hg.1 | ENST00000451152.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900151140.hg.1 | ENST00000451596.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900151140.hg.1 | NR_110953.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900151140.hg.1 | ENST00000451152.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900151140.hg.1 | ENST00000451596.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900151140.hg.1 | NR_110953.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900151140.hg.1 | TC0900007556.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900151151.hg.1 | ENST00000436054.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900151151.hg.1 | NR_110952.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900151151.hg.1 | ENST00000436054.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900151151.hg.1 | NR_110952.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900151151.hg.1 | TC0900007556.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900151131.hg.1 | ENST00000436054.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900151131.hg.1 | ENST00000451152.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900151131.hg.1 | ENST00000451596.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900151131.hg.1 | NR_110952.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900151131.hg.1 | NR_110953.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900151131.hg.1 | ENST00000436054.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900151131.hg.1 | ENST00000451152.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900151131.hg.1 | ENST00000451596.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900151131.hg.1 | NR_110952.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900151131.hg.1 | NR_110953.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900151131.hg.1 | TC0900007556.hg.1 | Clariom D TC |
