Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011356.hg.1 |
ENST00000421166.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011356.hg.1 |
ENST00000421166.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011356.hg.1 |
NR_110983.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011356.hg.1 |
NR_110983.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011356.hg.1 |
NR_110983.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011356.hg.1 |
NR_110983.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011356.hg.1 |
NR_110983.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011356.hg.1 |
NR_110984.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011356.hg.1 |
NR_110984.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011356.hg.1 |
NR_110984.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011356.hg.1 |
NR_110984.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011356.hg.1 |
NR_110984.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011356.hg.1 |
ENST00000421166.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011356.hg.1 |
ENST00000421166.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011356.hg.1 |
NR_110983.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011356.hg.1 |
NR_110983.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011356.hg.1 |
NR_110983.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011356.hg.1 |
NR_110983.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011356.hg.1 |
NR_110983.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011356.hg.1 |
NR_110984.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011356.hg.1 |
NR_110984.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011356.hg.1 |
NR_110984.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011356.hg.1 |
NR_110984.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011356.hg.1 |
NR_110984.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100183445.hg.1 |
ENST00000421166.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100183445.hg.1 |
NR_110983.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100183445.hg.1 |
NR_110984.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100183445.hg.1 |
ENST00000421166.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100183445.hg.1 |
NR_110983.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100183445.hg.1 |
NR_110984.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100183445.hg.1 |
TC0100011356.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100183449.hg.1 |
NR_110983.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100183449.hg.1 |
NR_110984.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100183449.hg.1 |
NR_110983.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100183449.hg.1 |
NR_110984.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100183449.hg.1 |
TC0100011356.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100183444.hg.1 |
ENST00000421166.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100183444.hg.1 |
NR_110983.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100183444.hg.1 |
ENST00000421166.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100183444.hg.1 |
NR_110983.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100183444.hg.1 |
TC0100011356.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100183448.hg.1 |
NR_110983.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100183448.hg.1 |
NR_110984.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100183448.hg.1 |
NR_110983.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100183448.hg.1 |
NR_110984.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100183448.hg.1 |
TC0100011356.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100183447.hg.1 |
NR_110983.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100183447.hg.1 |
NR_110984.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100183447.hg.1 |
NR_110983.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100183447.hg.1 |
NR_110984.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100183447.hg.1 |
TC0100011356.hg.1 |
Clariom D TC |