Assay Details
| Gene Symbol: DEPDC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110988.1 | 33 - 34 | 3599 | 93 | ||
| XM_011530565.2 | XP_011528867.1 | 36 - 37 | 3906 | 93 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | proline rich 14 like | PRR14L | C22orf30 | -124.1kb | Anti-sense | Chr. 22 - 31681347 - 31750176 on NCBI Build GRCh38 |
| Downstream | chromosome 22 open reading frame 24 | C22orf24 | HSN44A4A | 45.6kb | Anti-sense | Chr. 22 - 31933521 - 31945362 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

DEPDC5
Genomic Map