Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007743.hg.1 |
ENST00000423515.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007743.hg.1 |
ENST00000423515.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007743.hg.1 |
ENST00000423515.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007743.hg.1 |
ENST00000423515.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007743.hg.1 |
ENST00000423515.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007743.hg.1 |
ENST00000423515.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007743.hg.1 |
ENST00000423515.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007743.hg.1 |
ENST00000423515.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007743.hg.1 |
ENST00000423515.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007743.hg.1 |
ENST00000423515.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007743.hg.1 |
NR_110995.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007743.hg.1 |
NR_110995.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007743.hg.1 |
NR_110995.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007743.hg.1 |
NR_110995.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007743.hg.1 |
NR_110995.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007743.hg.1 |
NR_110995.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007743.hg.1 |
NR_110995.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007743.hg.1 |
NR_110995.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007743.hg.1 |
NR_110995.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007743.hg.1 |
NR_110995.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900152354.hg.1 |
ENST00000423515.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900152354.hg.1 |
NR_110995.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900152354.hg.1 |
ENST00000423515.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900152354.hg.1 |
NR_110995.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900152354.hg.1 |
TC0900007743.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900152355.hg.1 |
ENST00000423515.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900152355.hg.1 |
NR_110995.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900152355.hg.1 |
ENST00000423515.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900152355.hg.1 |
NR_110995.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900152355.hg.1 |
TC0900007743.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900007743.hg.1 |
ENST00000423515.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007743.hg.1 |
ENST00000423515.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007743.hg.1 |
ENST00000423515.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007743.hg.1 |
ENST00000423515.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007743.hg.1 |
ENST00000423515.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007743.hg.1 |
ENST00000423515.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007743.hg.1 |
ENST00000423515.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007743.hg.1 |
ENST00000423515.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007743.hg.1 |
ENST00000423515.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007743.hg.1 |
ENST00000423515.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007743.hg.1 |
NR_110995.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007743.hg.1 |
NR_110995.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007743.hg.1 |
NR_110995.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007743.hg.1 |
NR_110995.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007743.hg.1 |
NR_110995.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007743.hg.1 |
NR_110995.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007743.hg.1 |
NR_110995.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007743.hg.1 |
NR_110995.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007743.hg.1 |
NR_110995.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007743.hg.1 |
NR_110995.1 |
Refseq |