Assay Details
| Gene Symbol: KIF5C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_111932.1 | 18 - 19 | 2297 | 102 | ||
| XM_017004062.1 | XP_016859551.1 | 25 - 26 | 3266 | 102 | ||
| GenBank mRNA | BC110287.1 | - | 18 - 19 | 2227 | 5165262 | 102 |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101928553 | LOC101928553 | -84.4kb | Anti-sense | Chr. 2 - 148875995 - 148890572 on NCBI Build GRCh38 | |
| Downstream | LY6/PLAUR domain containing 6B | LYPD6B |
CT116, LYPD7 |
18.1kb | Sense | Chr. 2 - 149038467 - 149215262 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
organelle organization
microtubule-based movement
axon guidance
motor neuron axon guidance
protein localization
cytoskeleton-dependent intracellular transport
mRNA transport
microtubule motor activity
protein binding
ATP binding
microtubule binding
ATP-dependent microtubule motor activity, plus-end-directed
apolipoprotein receptor binding
organelle organization
microtubule-based movement
axon guidance
motor neuron axon guidance
protein localization
cytoskeleton-dependent intracellular transport
mRNA transport
microtubule motor activity
protein binding
ATP binding
microtubule binding
ATP-dependent microtubule motor activity, plus-end-directed
apolipoprotein receptor binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000450621.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000450621.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000450621.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000450621.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200167268.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167268.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200167268.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167268.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200167268.hg.1 | TC0200009616.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000450621.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000450621.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000450621.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000450621.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |

KIF5C
Genomic Map